Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 2 - 3 | 1032 | 71 | |
GenBank mRNA | AF083341.1 | - | 2 - 3 | 468 | 71 | |
AF135265.1 | - | 2 - 3 | 886 | 71 | ||
AY330290.1 | - | 2 - 3 | 357 | 71 | ||
AY330292.1 | - | 2 - 3 | 357 | 71 | ||
AY330293.2 | - | 2 - 3 | 357 | 71 | ||
AY330294.1 | - | 2 - 3 | 357 | 71 | ||
AY344964.1 | - | 2 - 3 | 357 | 71 | ||
AY344965.2 | - | 2 - 3 | 861 | 71 | ||
U40603.1 | - | 2 - 3 | 1032 | 71 | ||
U55995.1 | - | 2 - 3 | 510 | 71 | ||
U93052.1 | - | 2 - 3 | 458 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |