Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133284.2 | NP_579818.1 | 2 - 3 | 276 | 82 | |
GenBank mRNA | X04644.1 | - | 2 - 3 | 276 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |