Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012801.1 | NP_036933.1 | 4 - 5 | 647 | 62 | |
XM_006248918.2 | XP_006248980.1 | 2 - 3 | 629 | 62 | ||
XM_006248919.2 | XP_006248981.1 | 2 - 3 | 629 | 62 | ||
XM_008768940.1 | XP_008767162.1 | 3 - 4 | 2228 | 62 | ||
GenBank mRNA | BC061731.1 | - | 2 - 3 | 634 | 5598667 | 62 |
D10106.1 | - | 2 - 3 | 206 | 62 | ||
L06238.1 | - | 4 - 5 | 647 | 62 | ||
L06894.1 | - | 3 - 4 | 291 | 62 | ||
Z14120.1 | - | 2 - 3 | 141 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |