Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181086.3 | NP_851600.1 | 1 - 2 | 123 | 77 | |
GenBank mRNA | AY255102.1 | - | 1 - 2 | 94 | 77 | |
BC060537.1 | - | 1 - 2 | 127 | 6887545 | 77 | |
FQ229590.1 | - | 1 - 2 | 123 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |