Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Niacr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181476.1 | NP_852141.1 | 1 | 553 | 102 | |
GenBank mRNA | AB103062.1 | - | 1 | 553 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |