Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023989.1 | NP_076479.1 | 6 - 7 | 646 | 86 | |
XM_008768805.1 | XP_008767027.1 | 6 - 7 | 777 | 86 | ||
GenBank mRNA | AF260129.1 | - | 6 - 7 | 646 | 86 | |
FQ210639.1 | - | 6 - 7 | 741 | 86 | ||
FQ212134.1 | - | 6 - 7 | 741 | 86 | ||
FQ222500.1 | - | 5 - 6 | 589 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |