Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031572.1 | NP_113760.1 | 4 - 5 | 641 | 147 | |
GenBank mRNA | BC089790.1 | - | 4 - 5 | 649 | 7371667 | 147 |
FQ219038.1 | - | 2 - 3 | 322 | 147 | ||
FQ219491.1 | - | 3 - 4 | 516 | 147 | ||
FQ219720.1 | - | 1 - 2 | 111 | 147 | ||
J03786.1 | - | 4 - 5 | 641 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |