Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012899.2 | NP_037031.1 | 10 - 11 | 966 | 84 | |
XM_006238234.2 | XP_006238296.1 | 10 - 11 | 1559 | 84 | ||
XM_006238235.1 | XP_006238297.1 | 10 - 11 | 1056 | 84 | ||
GenBank mRNA | BC061806.1 | - | 10 - 11 | 966 | 5598340 | 84 |
FQ210250.1 | - | 10 - 11 | 911 | 84 | ||
FQ213585.1 | - | 10 - 11 | 1108 | 84 | ||
FQ218546.1 | - | 10 - 11 | 911 | 84 | ||
FQ231154.1 | - | 10 - 11 | 942 | 84 | ||
FQ232546.1 | - | 10 - 11 | 942 | 84 | ||
FQ233724.1 | - | 10 - 11 | 992 | 84 | ||
FQ234245.1 | - | 10 - 11 | 942 | 84 | ||
FQ234573.1 | - | 10 - 11 | 990 | 84 | ||
X04959.1 | - | 9 - 10 | 848 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |