Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058485.1 | - | 106 | 6921326 | 67 | |
FQ211494.1 | - | 106 | 67 | |||
FQ211970.1 | - | 102 | 67 | |||
FQ212357.1 | - | 103 | 67 | |||
FQ212579.1 | - | 102 | 67 | |||
FQ212640.1 | - | 102 | 67 | |||
FQ212789.1 | - | 106 | 67 | |||
FQ212869.1 | - | 106 | 67 | |||
FQ213028.1 | - | 102 | 67 | |||
FQ213029.1 | - | 100 | 67 | |||
FQ213091.1 | - | 102 | 67 | |||
FQ213115.1 | - | 108 | 67 | |||
FQ213216.1 | - | 102 | 67 | |||
FQ213366.1 | - | 101 | 67 | |||
FQ213496.1 | - | 105 | 67 | |||
FQ213520.1 | - | 106 | 67 | |||
FQ213526.1 | - | 102 | 67 | |||
FQ213581.1 | - | 102 | 67 | |||
FQ213598.1 | - | 102 | 67 | |||
FQ213637.1 | - | 102 | 67 | |||
FQ213695.1 | - | 102 | 67 | |||
FQ213703.1 | - | 102 | 67 | |||
FQ213760.1 | - | 102 | 67 | |||
FQ213762.1 | - | 107 | 67 | |||
FQ213913.1 | - | 102 | 67 | |||
FQ213941.1 | - | 106 | 67 | |||
FQ213981.1 | - | 103 | 67 | |||
FQ214059.1 | - | 106 | 67 | |||
FQ214181.1 | - | 106 | 67 | |||
FQ214262.1 | - | 106 | 67 | |||
FQ214491.1 | - | 102 | 67 | |||
FQ214500.1 | - | 102 | 67 | |||
FQ214519.1 | - | 102 | 67 | |||
FQ214540.1 | - | 106 | 67 | |||
FQ215317.1 | - | 102 | 67 | |||
FQ215600.1 | - | 106 | 67 | |||
FQ216397.1 | - | 102 | 67 | |||
FQ220250.1 | - | 102 | 67 | |||
FQ220325.1 | - | 102 | 67 | |||
FQ220698.1 | - | 106 | 67 | |||
FQ220779.1 | - | 102 | 67 | |||
FQ222184.1 | - | 105 | 67 | |||
FQ223357.1 | - | 106 | 67 | |||
FQ226994.1 | - | 106 | 67 | |||
FQ229228.1 | - | 102 | 67 | |||
FQ229580.1 | - | 106 | 67 | |||
FQ229619.1 | - | 102 | 67 | |||
FQ229790.1 | - | 102 | 67 | |||
FQ231022.1 | - | 106 | 67 | |||
FQ233337.1 | - | 105 | 67 | |||
FQ234377.1 | - | 106 | 67 | |||
FQ235266.1 | - | 103 | 67 | |||
M17069.1 | - | 104 | 67 | |||
M19312.1 | - | 85 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |