Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053884.1 | NP_446336.1 | 1 - 2 | 208 | 72 | |
GenBank mRNA | U43175.1 | - | 1 - 2 | 208 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |