Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105982.1 | NP_001099452.1 | 1 - 2 | 140 | 56 | |
XM_006250373.2 | XP_006250435.1 | 1 - 2 | 294 | 56 | ||
XM_008769820.1 | XP_008768042.1 | 1 - 2 | 294 | 56 | ||
XM_008769823.1 | XP_008768045.1 | 1 - 2 | 294 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |