Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012146.1 | NP_001012146.1 | 8 - 9 | 1425 | 55 | |
XM_008768890.1 | XP_008767112.1 | 8 - 9 | 1408 | 55 | ||
GenBank mRNA | BC081905.1 | - | 8 - 9 | 1425 | 7111034 | 55 |
FQ211720.1 | - | 8 - 9 | 1349 | 55 | ||
FQ228410.1 | - | 8 - 9 | 1327 | 55 |
Gene Symbol: LOC100910275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750241.2 | XP_003750289.1 | 8 - 9 | 1424 | 55 | |
XM_006240675.2 | XP_006240737.1 | 8 - 9 | 1401 | 55 |
Gene Symbol: LOC103693608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768936.1 | XP_008767158.1 | 2 - 3 | 548 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |