Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031693.2 | NP_113881.1 | 3 - 4 | 1204 | 65 | |
GenBank mRNA | DQ181551.1 | - | 3 - 4 | 968 | 65 | |
FQ211592.1 | - | 3 - 4 | 1201 | 65 | ||
L38247.1 | - | 3 - 4 | 1205 | 65 | ||
U14398.1 | - | 3 - 4 | 1233 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |