Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankdd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243300.2 | XP_006243362.1 | 13 - 14 | 2628 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |