Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100750.1 | NP_001094220.1 | 5 - 6 | 577 | 70 | |
GenBank mRNA | BC079367.1 | - | 5 - 6 | 570 | 70 | |
BC089863.1 | - | 4 - 5 | 435 | 70 | ||
BC099746.1 | - | 5 - 6 | 561 | 70 | ||
BC160886.1 | - | 5 - 6 | 577 | 7385389 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |