Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109249.1 | NP_001102719.1 | 2 - 3 | 528 | 85 | |
XM_006236864.2 | XP_006236926.1 | 2 - 3 | 542 | 85 | ||
XM_008763079.1 | XP_008761301.1 | 2 - 3 | 543 | 85 | ||
GenBank mRNA | BC168694.1 | - | 2 - 3 | 505 | 7937199 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |