Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770413.1 | XP_008768635.1 | 4 - 5 | 666 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |