Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010953.1 | NP_001010953.1 | 1 - 2 | 248 | 70 | |
GenBank mRNA | BC089058.1 | - | 1 - 2 | 248 | 7315480 | 70 |
D82867.1 | - | 1 - 2 | 278 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |