Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271132.1 | NP_001258061.1 | 2 - 3 | 4378 | 92 | |
XM_006228412.2 | XP_006228474.1 | 2 - 3 | 4943 | 92 | ||
XM_008758918.1 | XP_008757140.1 | 3 - 4 | 4203 | 92 | ||
XM_008758919.1 | XP_008757141.1 | 2 - 3 | 3996 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |