Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077429.1 | NP_001070897.1 | 3 - 4 | 188 | 107 | |
XM_006234377.2 | XP_006234439.1 | 3 - 4 | 270 | 107 | ||
XM_006234378.2 | XP_006234440.1 | 4 - 5 | 291 | 107 | ||
XM_006234379.2 | XP_006234441.1 | 3 - 4 | 318 | 107 | ||
XM_006234380.2 | XP_006234442.1 | 3 - 4 | 390 | 107 | ||
XM_006234381.2 | XP_006234443.1 | 3 - 4 | 483 | 107 | ||
XM_006234382.2 | XP_006234444.1 | 4 - 5 | 411 | 107 | ||
XM_006234385.2 | XP_006234447.1 | 3 - 4 | 272 | 107 | ||
XM_008761909.1 | XP_008760131.1 | 3 - 4 | 1367 | 107 | ||
XM_008761910.1 | XP_008760132.1 | 3 - 4 | 272 | 107 | ||
GenBank mRNA | BC126100.1 | - | 3 - 4 | 188 | 7132654 | 107 |
FQ218047.1 | - | 2 - 3 | 67 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |