Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234794.2 | XP_006234856.1 | 6 - 7 | 2646 | 95 | |
XM_006234796.2 | XP_006234858.1 | 6 - 7 | 4447 | 95 | ||
XM_008762115.1 | XP_008760337.1 | 6 - 7 | 2316 | 95 | ||
XM_008762116.1 | XP_008760338.1 | 6 - 7 | 2313 | 95 | ||
XM_008762117.1 | XP_008760339.1 | 6 - 7 | 2307 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |