Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105717.2 | NP_001099187.1 | 4 - 5 | 697 | 70 | |
XM_006252097.2 | XP_006252159.1 | 4 - 5 | 928 | 70 | ||
GenBank mRNA | CB729753.1 | - | 1 - 2 | 103 | 70 | |
Z46882.1 | - | 4 - 5 | 650 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |