Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024748.1 | NP_001019919.1 | 1 - 2 | 685 | 86 | |
XM_008758909.1 | XP_008757131.1 | 1 - 2 | 740 | 86 | ||
GenBank mRNA | BC097421.1 | - | 1 - 2 | 685 | 7454827 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |