Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025731.1 | NP_001020902.1 | 5 - 6 | 702 | 74 | |
XM_006233836.2 | XP_006233898.1 | 5 - 6 | 738 | 74 | ||
XM_006233838.2 | XP_006233900.1 | 5 - 6 | 737 | 74 | ||
GenBank mRNA | BC099088.1 | - | 5 - 6 | 702 | 7456179 | 74 |
FQ217346.1 | - | 5 - 6 | 723 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |