Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107831.3 | NP_001101301.2 | 3 - 4 | 452 | 115 | |
GenBank mRNA | BC168897.1 | - | 3 - 4 | 391 | 8362491 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |