Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057141.1 | NP_476482.1 | 13 - 14 | 1220 | 75 | |
XM_006253552.2 | XP_006253614.1 | 15 - 16 | 1627 | 75 | ||
XM_006253553.2 | XP_006253615.1 | 14 - 15 | 1323 | 75 | ||
XM_006253554.2 | XP_006253616.1 | 15 - 16 | 1627 | 75 | ||
XM_006253555.2 | XP_006253617.1 | 14 - 15 | 1555 | 75 | ||
GenBank mRNA | BC061867.1 | - | 14 - 15 | 1307 | 5598872 | 75 |
D17711.1 | - | 13 - 14 | 1220 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |