Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031534.2 | NP_113722.2 | 9 - 10 | 1247 | 57 | |
XM_006234620.1 | XP_006234682.1 | 8 - 9 | 1589 | 57 | ||
GenBank mRNA | CO382418.1 | - | 1 - 2 | 115 | 57 | |
X69716.1 | - | 9 - 10 | 1247 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |