Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105898.1 | NP_001099368.1 | 2 - 3 | 329 | 81 | |
XM_006247994.2 | XP_006248056.1 | 2 - 3 | 370 | 81 | ||
XM_006247995.2 | XP_006248057.1 | 2 - 3 | 370 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |