Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031628.1 | NP_113816.1 | 7 - 8 | 2337 | 67 | |
XM_008763702.1 | XP_008761924.1 | 8 - 9 | 4131 | 67 | ||
XM_008763703.1 | XP_008761925.1 | 7 - 8 | 4059 | 67 | ||
GenBank mRNA | D38530.1 | - | 7 - 8 | 2337 | 67 | |
DQ268830.1 | - | 7 - 8 | 2337 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |