Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007144.1 | NP_001007145.1 | 2 - 3 | 109 | 72 | |
XM_006238360.2 | XP_006238422.1 | 2 - 3 | 145 | 72 | ||
XM_008763778.1 | XP_008762000.1 | 2 - 3 | 145 | 72 | ||
GenBank mRNA | BC085861.1 | - | 2 - 3 | 109 | 7190361 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |