Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108436.1 | NP_001101906.1 | 4 - 5 | 635 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |