Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022245.1 | NP_071581.1 | 2 - 3 | 327 | 79 | |
GenBank mRNA | AF007108.1 | - | 2 - 3 | 290 | 79 | |
BC086945.1 | - | 2 - 3 | 321 | 7322141 | 79 | |
D13205.1 | - | 2 - 3 | 327 | 79 | ||
FQ209861.1 | - | 2 - 3 | 318 | 79 | ||
FQ210790.1 | - | 2 - 3 | 315 | 79 | ||
FQ224354.1 | - | 2 - 3 | 318 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |