Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106224.2 | NP_001099694.1 | 3 - 4 | 472 | 58 | |
GenBank mRNA | BC169077.1 | - | 3 - 4 | 472 | 5620985 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |