Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106920.1 | NP_001100390.1 | 8 - 9 | 1992 | 87 | |
XM_006245167.2 | XP_006245229.1 | 8 - 9 | 1005 | 87 | ||
XM_006245168.2 | XP_006245230.1 | 8 - 9 | 1171 | 87 | ||
XM_006245169.2 | XP_006245231.1 | 8 - 9 | 1168 | 87 | ||
GenBank mRNA | BC159424.1 | - | 8 - 9 | 956 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |