Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024891.1 | NP_001020062.1 | 1 - 2 | 194 | 121 | |
GenBank mRNA | BC097416.1 | - | 1 - 2 | 194 | 7454359 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |