Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007713.1 | NP_001007714.1 | 7 - 8 | 601 | 84 | |
XM_008767238.1 | XP_008765460.1 | 7 - 8 | 2973 | 84 | ||
XM_008767239.1 | XP_008765461.1 | 8 - 9 | 624 | 84 | ||
GenBank mRNA | BC079087.1 | - | 7 - 8 | 601 | 7123233 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |