Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhej1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014217.1 | NP_001014239.1 | 2 - 3 | 281 | 110 | |
XM_006245240.1 | XP_006245302.1 | 2 - 3 | 359 | 110 | ||
XM_006245241.2 | XP_006245303.1 | 2 - 3 | 935 | 110 | ||
XM_006245242.2 | XP_006245304.1 | 2 - 3 | 704 | 110 | ||
GenBank mRNA | BC079033.1 | - | 2 - 3 | 281 | 7114269 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |