Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019310.1 | NP_062183.1 | 1 | 872 | 58 | |
XM_006245273.2 | XP_006245335.2 | 2 | 949 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |