Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106916.1 | NP_001100386.1 | 1 - 2 | 139 | 84 | |
XM_008767217.1 | XP_008765439.1 | 1 - 2 | 134 | 84 | ||
XM_008767219.1 | XP_008765441.1 | 1 - 2 | 149 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |