Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hesx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109106.1 | NP_001102576.1 | 2 - 3 | 715 | 77 | |
XM_008771001.1 | XP_008769223.1 | 4 - 5 | 811 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |