Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031336.1 | NP_112626.1 | 1 - 2 | 836 | 65 | |
GenBank mRNA | AB017820.1 | - | 1 - 2 | 836 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |