Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144741.1 | NP_653342.1 | 1 - 2 | 72 | 100 | |
GenBank mRNA | AF378366.1 | - | 1 - 2 | 72 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |