Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106847.2 | NP_001100317.2 | 3 - 4 | 315 | 94 | |
XM_006243556.2 | XP_006243618.1 | 616 | 94 | |||
XM_006243558.2 | XP_006243620.1 | 3 - 4 | 1715 | 94 | ||
XM_006243561.2 | XP_006243623.1 | 2 - 3 | 1675 | 94 | ||
XM_006243572.2 | XP_006243634.1 | 2 - 3 | 1671 | 94 | ||
XM_008766479.1 | XP_008764701.1 | 4 - 5 | 294 | 94 | ||
XM_008766480.1 | XP_008764702.1 | 4 - 5 | 293 | 94 | ||
XM_008766481.1 | XP_008764703.1 | 4 - 5 | 294 | 94 | ||
XM_008766487.1 | XP_008764709.1 | 5 - 6 | 423 | 94 | ||
XM_008766488.1 | XP_008764710.1 | 2 - 3 | 130 | 94 | ||
GenBank mRNA | BC168961.1 | - | 3 - 4 | 315 | 7444676 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |