Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112737.2 | NP_001106208.1 | 1 - 2 | 1759 | 59 | |
GenBank mRNA | BC168666.1 | - | 1 - 2 | 1759 | 7933386 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |