Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126048.2 | NP_001119520.1 | 1 - 2 | 87 | 66 | |
GenBank mRNA | BC157813.1 | - | 1 - 2 | 87 | 66 | |
FQ221914.1 | - | 1 - 2 | 63 | 66 | ||
FQ222972.1 | - | 1 - 2 | 52 | 66 |
Gene Symbol: LOC100910678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749743.3 | XP_003749791.1 | 1 - 2 | 63 | 66 | |
XM_006236476.2 | XP_006236538.1 | 1 | 65 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |