Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108323.1 | NP_001101793.1 | 4 - 5 | 402 | 75 | |
XM_006248628.2 | XP_006248690.1 | 4 - 5 | 527 | 75 | ||
XM_006248629.2 | XP_006248691.1 | 4 - 5 | 397 | 75 | ||
XM_006248630.2 | XP_006248692.1 | 4 - 5 | 422 | 75 | ||
XM_006248632.2 | XP_006248694.1 | 4 - 5 | 527 | 75 | ||
XM_006248633.2 | XP_006248695.1 | 4 - 5 | 526 | 75 | ||
XM_006248634.2 | XP_006248696.1 | 4 - 5 | 528 | 75 | ||
XM_008768888.1 | XP_008767110.1 | 4 - 5 | 400 | 75 | ||
XM_008768889.1 | XP_008767111.1 | 4 - 5 | 527 | 75 | ||
GenBank mRNA | FQ213583.1 | - | 3 - 4 | 429 | 75 | |
FQ217948.1 | - | 4 - 5 | 529 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |