Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107078.2 | NP_001100548.1 | 4 - 5 | 1778 | 64 | |
XM_006248693.2 | XP_006248755.1 | 5 - 6 | 2151 | 64 | ||
XM_006248694.2 | XP_006248756.1 | 5 - 6 | 1971 | 64 | ||
XM_006248695.2 | XP_006248757.1 | 5 - 6 | 2148 | 64 | ||
XM_006248696.2 | XP_006248758.1 | 4 - 5 | 2006 | 64 | ||
XM_006248697.2 | XP_006248759.1 | 4 - 5 | 2003 | 64 | ||
XM_006248698.2 | XP_006248760.1 | 4 - 5 | 1855 | 64 | ||
GenBank mRNA | BC169079.1 | - | 4 - 5 | 1778 | 7937326 | 64 |
FQ214252.1 | - | 4 - 5 | 1972 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |