Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039021.1 | NP_001034110.1 | 2 - 3 | 358 | 61 | |
XM_006248610.2 | XP_006248672.1 | 2 - 3 | 357 | 61 | ||
GenBank mRNA | AY871204.1 | - | 2 - 3 | 358 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |