Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035249.1 | NP_001030326.1 | 8 - 9 | 966 | 68 | |
GenBank mRNA | BC088226.1 | - | 8 - 9 | 884 | 68 | |
BC105907.1 | - | 8 - 9 | 966 | 68 | ||
BC126098.1 | - | 8 - 9 | 998 | 68 | ||
BC158564.1 | - | 8 - 9 | 950 | 8374563 | 68 | |
FQ214115.1 | - | 8 - 9 | 961 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |