Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053942.1 | NP_446394.1 | 1 | 600 | 156 | |
GenBank mRNA | AB113391.1 | - | 1 | 600 | 156 | |
AF177688.1 | - | 1 | 214 | 156 | ||
AY573982.1 | - | 1 | 600 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |